Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205938.1 | NP_001192867.1 | 12 - 13 | 2182 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |