Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205969.1 | NP_001192898.1 | 14 - 15 | 1716 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |