Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206011.1 | NP_001192940.1 | 5 - 6 | 1394 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |