Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206016.1 | NP_001192945.1 | 4 - 5 | 1168 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |