Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206026.1 | NP_001192955.1 | 7 - 8 | 952 | 65 | |
GenBank mRNA | AY143368.1 | - | 7 - 8 | 914 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |