Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206026.1 | NP_001192955.1 | 9 - 10 | 1197 | 104 | |
GenBank mRNA | AY143368.1 | - | 9 - 10 | 1159 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |