Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEFTY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206043.2 | NP_001192972.2 | 1 - 2 | 249 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |