Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206075.1 | NP_001193004.1 | 7 - 8 | 674 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |