Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206094.1 | NP_001193023.1 | 4 - 5 | 837 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |