Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206140.1 | NP_001193069.1 | 3 - 4 | 371 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |