Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206171.3 | NP_001193100.1 | 3 - 4 | 1716 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |