Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206180.1 | NP_001193109.1 | 4 - 5 | 503 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |