Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206246.2 | NP_001193175.1 | 4 - 5 | 742 | 79 | |
GenBank mRNA | AJ627254.1 | - | 3 - 4 | 500 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |