Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206246.2 | NP_001193175.1 | 7 - 8 | 1148 | 71 | |
GenBank mRNA | AJ627254.1 | - | 6 - 7 | 906 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |