Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206253.1 | NP_001193182.1 | 6 - 7 | 753 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |