Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206259.1 | NP_001193188.1 | 4 - 5 | 439 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |