Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192467.1 | NP_001179396.1 | 1 - 2 | 163 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |