Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192467.1 | NP_001179396.1 | 7 - 8 | 942 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |