Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192532.2 | NP_001179461.2 | 10 - 11 | 1115 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |