Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192534.1 | NP_001179463.1 | 4 - 5 | 710 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |