Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192567.1 | NP_001179496.1 | 5 - 6 | 911 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |