Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192604.3 | NP_001179533.3 | 3 - 4 | 595 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |