Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192618.1 | NP_001179547.1 | 2 - 3 | 112 | 92 | |
GenBank mRNA | AY962103.1 | - | 2 - 3 | 72 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |