Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192650.1 | NP_001179579.1 | 4 - 5 | 636 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |