Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192657.2 | NP_001179586.2 | 10 - 11 | 1306 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |