Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192659.2 | NP_001179588.1 | 3 - 4 | 913 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Bos taurus Probe spans exons |