Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206282.3 | NP_001193211.1 | 2 - 3 | 1812 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |