Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206301.1 | NP_001193230.1 | 4 - 5 | 440 | 65 | |
GenBank mRNA | BC104541.1 | - | 3 - 4 | 422 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |