Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206325.2 | NP_001193254.1 | 7 - 8 | 1517 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |