Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206342.1 | NP_001193271.1 | 4 - 5 | 681 | 130 | |
GenBank mRNA | BC149797.1 | - | 4 - 5 | 681 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |