Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206342.1 | NP_001193271.1 | 6 - 7 | 963 | 125 | |
GenBank mRNA | BC149797.1 | - | 6 - 7 | 963 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |