Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206345.1 | NP_001193274.1 | 4 - 5 | 451 | 71 | |
GenBank mRNA | BC148886.1 | - | 4 - 5 | 386 | 71 | |
BT026295.1 | - | 4 - 5 | 454 | 71 | ||
DV837947.1 | - | 4 - 5 | 452 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |