Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206386.1 | NP_001193315.1 | 3 - 4 | 296 | 71 | |
GenBank mRNA | BC149586.1 | - | 298 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |