Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206387.1 | NP_001193316.1 | 4 - 5 | 417 | 87 | |
GenBank mRNA | BC111290.1 | - | 4 - 5 | 417 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |