Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206405.1 | NP_001193334.1 | 11 - 12 | 1673 | 64 | |
GenBank mRNA | BC103457.1 | - | 11 - 12 | 1663 | 64 | |
BC149976.1 | - | 11 - 12 | 1673 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |