Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206440.1 | NP_001193369.1 | 5 - 6 | 825 | 78 | |
GenBank mRNA | BC151821.1 | - | 5 - 6 | 826 | 78 | |
CX954191.1 | - | 1 - 2 | 278 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |