Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206470.1 | NP_001193399.1 | 6 - 7 | 908 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |