Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206509.1 | NP_001193438.1 | 3 - 4 | 402 | 93 | |
GenBank mRNA | EV658200.1 | - | 3 - 4 | 404 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |