Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206545.1 | NP_001193474.1 | 1 - 2 | 310 | 112 | |
GenBank mRNA | BE749356.1 | - | 1 - 2 | 310 | 112 | |
MG934429.1 | - | 1 - 2 | 332 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Bos taurus Probe spans exons |