Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206564.1 | NP_001193493.1 | 6 - 7 | 496 | 96 | |
GenBank mRNA | BC110167.1 | - | 7 - 8 | 870 | 96 | |
DV869571.1 | - | 6 - 7 | 482 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |