Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192678.1 | NP_001179607.1 | 2 - 3 | 356 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |