Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192714.2 | NP_001179643.1 | 5 - 6 | 1112 | 89 | |
GenBank mRNA | DQ072112.1 | - | 1 - 2 | 74 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |