Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192722.2 | NP_001179651.1 | 4 - 5 | 861 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |