Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192736.1 | NP_001179665.1 | 5 - 6 | 903 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |