Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192782.3 | NP_001179711.2 | 6 - 7 | 4636 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |