Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192793.2 | NP_001179722.1 | 3 - 4 | 706 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |