Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192801.1 | NP_001179730.1 | 6 - 7 | 1035 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |