Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192804.1 | NP_001179733.1 | 3 - 4 | 919 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |