Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192813.1 | NP_001179742.1 | 1 - 2 | 1058 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |