Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192820.3 | NP_001179749.1 | 3 - 4 | 280 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |